Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sym-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078025.7 | NP_510426.1 | 5 - 6 | 1266 | 78 | |
GenBank mRNA | AJ242473.1 | - | 5 - 6 | 1170 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |