Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ent-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268644.2 | NP_001255573.1 | 3 - 4 | 431 | 173 | |
GenBank mRNA | AF435979.1 | - | 3 - 4 | 425 | 173 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |