Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ent-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268644.2 | NP_001255573.1 | 4 - 5 | 942 | 152 | |
GenBank mRNA | AF435979.1 | - | 4 - 5 | 936 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |