Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nspd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059358.6 | NP_491759.1 | 1 | 154 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Both primers and probe map within a single exon |