Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bath-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062111.5 | NP_494512.2 | 2 - 3 | 527 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |