Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027880.4 | NP_001023051.1 | 2 | 57 | 61 | |
GenBank mRNA | U38935.1 | - | 2 | 105 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Both primers and probe map within a single exon |