Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028399.4 | NP_001023570.2 | 9 - 10 | 1511 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |