Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fce-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028776.3 | NP_001023947.1 | 5 - 6 | 480 | 66 | |
GenBank mRNA | AJ487543.1 | - | 5 - 6 | 502 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |