Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sri-67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269962.1 | NP_001256891.1 | 1 - 2 | 156 | 109 | |
NM_001269963.1 | NP_001256892.1 | 1 - 2 | 156 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |