Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: far-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073849.6 | NP_506250.2 | 1 - 2 | 458 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |