Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adpr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026658.2 | NP_001021829.1 | 6 - 7 | 1221 | 73 | |
NM_001306479.3 | NP_001293408.1 | 3 - 4 | 597 | 73 | ||
NM_001306480.3 | NP_001293409.1 | 2 - 3 | 444 | 73 | ||
NM_001306481.3 | NP_001293410.1 | 1 - 2 | 96 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |