Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026805.2 | NP_001021976.1 | 7 - 8 | 893 | 86 | |
GenBank mRNA | DQ015968.1 | - | 7 - 8 | 815 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |