Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pac-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136361.5 | NP_001129833.1 | 2 - 3 | 400 | 112 | |
NM_001306728.4 | NP_001293657.1 | 2 - 3 | 410 | 112 | ||
NM_066476.7 | NP_498877.4 | 2 - 3 | 308 | 112 | ||
GenBank mRNA | EU752496.1 | - | 2 - 3 | 400 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |