Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268567.3 | NP_001255496.1 | 3 - 4 | 568 | 141 | |
NM_001268568.3 | NP_001255497.1 | 2 - 3 | 234 | 141 | ||
NR_131602.1 | 3 - 4 | 339 | 141 | |||
NR_131603.1 | 3 - 4 | 290 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |