Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268567.3 | NP_001255496.1 | 5 - 6 | 815 | 66 | |
NM_001268568.3 | NP_001255497.1 | 4 - 5 | 481 | 66 | ||
NR_131602.1 | 5 - 6 | 586 | 66 | |||
NR_131603.1 | 5 - 6 | 537 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |