Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsx-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074376.2 | NP_506777.2 | 3 - 4 | 319 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |