Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038424.4 | NP_001033513.2 | 8 - 9 | 1000 | 79 | |
NM_001330830.1 | NP_001317792.1 | 8 - 9 | 954 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |