Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624275.3 | XP_005624332.1 | 251 | 123 | ||
XM_005624276.3 | XP_005624333.1 | 251 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris |