Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031635.2 | NP_001026805.1 | 3 - 4 | 605 | 90 | |
XM_005623202.3 | XP_005623259.1 | 3 - 4 | 648 | 90 | ||
GenBank mRNA | DN368512.1 | - | 1 - 2 | 87 | 90 | |
DQ116955.1 | - | 1 - 2 | 439 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Inventoried Probe spans exons |