Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003160.1 | NP_001003160.1 | 4 - 5 | 623 | 98 | |
GenBank mRNA | AB021930.1 | - | 4 - 5 | 623 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |