Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003215.2 | NP_001003215.2 | 10 - 11 | 1222 | 138 | |
XM_014118873.2 | XP_013974348.1 | 11 - 12 | 1395 | 138 | ||
XM_014118874.2 | XP_013974349.1 | 11 - 12 | 1325 | 138 | ||
GenBank mRNA | AB066299.1 | - | 4 - 5 | 469 | 138 | |
AF045016.1 | - | 11 - 12 | 1288 | 138 | ||
AF536758.1 | - | 10 - 11 | 1238 | 138 | ||
AY582533.1 | - | 10 - 11 | 1222 | 138 | ||
DQ068953.1 | - | 10 - 11 | 1222 | 138 | ||
FJ617477.1 | - | 10 - 11 | 1222 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |