Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003215.2 | NP_001003215.2 | 12 - 13 | 1557 | 78 | |
XM_014118873.2 | XP_013974348.1 | 13 - 14 | 1730 | 78 | ||
XM_014118874.2 | XP_013974349.1 | 13 - 14 | 1660 | 78 | ||
GenBank mRNA | AB066299.1 | - | 6 - 7 | 804 | 78 | |
AF045016.1 | - | 13 - 14 | 1623 | 78 | ||
AF536758.1 | - | 12 - 13 | 1573 | 78 | ||
AJ419568.1 | - | 12 - 13 | 1559 | 78 | ||
AY582533.1 | - | 12 - 13 | 1557 | 78 | ||
DQ068953.1 | - | 12 - 13 | 1557 | 78 | ||
FJ617477.1 | - | 12 - 13 | 1557 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |