Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005263.1 | NP_001005263.1 | 8 - 9 | 1361 | 86 | |
GenBank mRNA | AB097057.1 | - | 8 - 9 | 1361 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |