Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AICDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003380.1 | NP_001003380.1 | 3 - 4 | 500 | 99 | |
XM_022410855.1 | XP_022266563.1 | 2 - 3 | 554 | 99 | ||
GenBank mRNA | AB122019.1 | - | 3 - 4 | 500 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |