Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOCS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031631.1 | NP_001026801.1 | 443 | 127 | ||
GenBank mRNA | AB164434.1 | - | 443 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |