Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006648.1 | NP_001006649.1 | 1 - 2 | 35 | 57 | |
XM_005640991.3 | XP_005641048.1 | 3 - 4 | 751 | 57 | ||
GenBank mRNA | AB193091.1 | - | 1 - 2 | 35 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Canis familiaris Probe spans exons |