Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003071.2 | NP_001003071.1 | 299 | 83 | ||
XM_005624324.3 | XP_005624381.1 | 497 | 83 | |||
XM_005624325.3 | XP_005624382.1 | 536 | 83 | |||
XM_005624326.2 | XP_005624383.1 | 384 | 83 | |||
XM_014116461.2 | XP_013971936.1 | 560 | 83 | |||
XM_014116462.2 | XP_013971937.1 | 554 | 83 | |||
XM_022422524.1 | XP_022278232.1 | 561 | 83 | |||
GenBank mRNA | AF038862.1 | - | 299 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris |