Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197044.1 | NP_001183973.1 | 2 - 3 | 338 | 74 | |
GenBank mRNA | AF049909.1 | - | 2 - 3 | 319 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |