Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003127.1 | NP_001003127.1 | 1196 | 86 | ||
GenBank mRNA | AF171061.1 | - | 1196 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris |