Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEKT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003097.2 | NP_001003097.2 | 2 - 3 | 386 | 85 | |
XM_014113168.2 | XP_013968643.1 | 2 - 3 | 347 | 85 | ||
XM_014113169.2 | XP_013968644.1 | 2 - 3 | 323 | 85 | ||
XM_014113170.2 | XP_013968645.1 | 2 - 3 | 225 | 85 | ||
XM_022418005.1 | XP_022273713.1 | 2 - 3 | 224 | 85 | ||
GenBank mRNA | AF191546.1 | - | 2 - 3 | 400 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons |