Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMPER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003197.2 | NP_001003197.2 | 660 | 65 | ||
GenBank mRNA | AF263546.2 | - | 660 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris |