Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003301.1 | NP_001003301.1 | 2 - 3 | 258 | 68 | |
GenBank mRNA | AF275796.1 | - | 2 - 3 | 201 | 68 | |
AF328929.1 | - | 2 - 3 | 199 | 68 | ||
U12234.1 | - | 2 - 3 | 258 | 68 | ||
Z70045.1 | - | 1 - 2 | 182 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Inventoried Probe spans exons |