Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003017.2 | NP_001003017.2 | 2 - 3 | 432 | 61 | |
XM_005622221.3 | XP_005622278.1 | 2 - 3 | 469 | 61 | ||
XM_022420725.1 | XP_022276433.1 | 2 - 3 | 472 | 61 | ||
GenBank mRNA | AY622224.1 | - | 2 - 3 | 432 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |