Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003017.2 | NP_001003017.2 | 8 - 9 | 1292 | 65 | |
XM_005622221.3 | XP_005622278.1 | 9 - 10 | 1485 | 65 | ||
GenBank mRNA | AY622224.1 | - | 8 - 9 | 1292 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |