Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003022.1 | NP_001003022.1 | 4 - 5 | 4328 | 82 | |
GenBank mRNA | AF483595.1 | - | 4 - 5 | 4328 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons |