Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003100.2 | NP_001003100.2 | 1 | 455 | 124 | |
GenBank mRNA | AJ388521.1 | - | 1 | 346 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |