Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002955.1 | NP_001002955.1 | 1 - 2 | 165 | 116 | |
GenBank mRNA | AJ388555.1 | - | 1 - 2 | 165 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |