Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195140.1 | NP_001182069.1 | 1 - 2 | 141 | 66 | |
GenBank mRNA | M85068.1 | - | 1 - 2 | 141 | 66 |
Gene Symbol: SLC5A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022413340.1 | XP_022269048.1 | 1 - 2 | 132 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |