Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003302.1 | NP_001003302.1 | 1 - 2 | 79 | 119 | |
GenBank mRNA | U15662.1 | - | 1 - 2 | 79 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |