Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025621.1 | NP_001020792.1 | 2 - 3 | 296 | 71 | |
GenBank mRNA | AM048627.1 | - | 2 - 3 | 296 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |