Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_532396.6 | XP_532396.1 | 4 - 5 | 520 | 78 | |
GenBank mRNA | AY004331.1 | - | 3 - 4 | 370 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Canis familiaris Inventoried Probe spans exons |