Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003064.2 | NP_001003064.2 | 4 - 5 | 620 | 89 | |
GenBank mRNA | AY062037.1 | - | 4 - 5 | 526 | 89 | |
DN365927.1 | - | 4 - 5 | 538 | 89 | ||
DN392813.1 | - | 4 - 5 | 629 | 89 | ||
DN871864.1 | - | 4 - 5 | 537 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |