Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003223.1 | NP_001003223.1 | 7 - 8 | 849 | 70 | |
XM_014114387.1 | XP_013969862.1 | 7 - 8 | 1130 | 70 | ||
XM_022419477.1 | XP_022275185.1 | 7 - 8 | 1060 | 70 | ||
GenBank mRNA | AY069922.1 | - | 7 - 8 | 825 | 70 | |
D29807.1 | - | 7 - 8 | 849 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Inventoried Probe spans exons |