Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002940.1 | NP_001002940.1 | 1 - 2 | 147 | 64 | |
GenBank mRNA | AY217754.1 | - | 1 - 2 | 147 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |