Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLANA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194968.1 | NP_001181897.1 | 2 - 3 | 169 | 85 | |
XM_005626364.3 | XP_005626421.1 | 6 - 7 | 891 | 85 | ||
XM_005626365.3 | XP_005626422.1 | 6 - 7 | 938 | 85 | ||
XM_005626366.2 | XP_005626423.1 | 6 - 7 | 818 | 85 | ||
XM_005626367.3 | XP_005626424.1 | 5 - 6 | 654 | 85 | ||
XM_005626368.3 | XP_005626425.1 | 4 - 5 | 572 | 85 | ||
XM_022424893.1 | XP_022280601.1 | 7 - 8 | 983 | 85 | ||
GenBank mRNA | AY251268.1 | - | 2 - 3 | 140 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |