Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003357.2 | NP_001003357.2 | 1 - 2 | 182 | 142 | |
GenBank mRNA | AY283372.1 | - | 1 - 2 | 33 | 142 | |
DN394227.1 | - | 1 - 2 | 192 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |