Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003364.1 | NP_001003364.1 | 8 - 9 | 1131 | 108 | |
GenBank mRNA | AY563036.1 | - | 8 - 9 | 1131 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |