Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005755.1 | NP_001005755.1 | 1 - 2 | 244 | 130 | |
XM_022427658.1 | XP_022283366.1 | 1 - 2 | 262 | 130 | ||
GenBank mRNA | AY725987.1 | - | 1 - 2 | 244 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |