Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024633.1 | NP_001019804.1 | 1 - 2 | 347 | 68 | |
GenBank mRNA | AY832859.1 | - | 1 - 2 | 347 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Canis familiaris Probe spans exons |