Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048016.1 | NP_001041481.1 | 13 - 14 | 1872 | 84 | |
XM_005626399.2 | XP_005626456.1 | 13 - 14 | 1707 | 84 | ||
XM_022424911.1 | XP_022280619.1 | 14 - 15 | 1785 | 84 | ||
GenBank mRNA | AY911512.1 | - | 13 - 14 | 1872 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |